Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94971

Supelco

Bisphenol P

analytical standard

Synonym(s):

4,4′-(1,4-Phenylenediisopropylidene)bisphenol, Bisphenol P

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[C(CH3)2C6H4OH]2
CAS Number:
Molecular Weight:
346.46
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

193-195 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(cc2)C(C)(C)c3ccc(O)cc3

InChI

1S/C24H26O2/c1-23(2,19-9-13-21(25)14-10-19)17-5-7-18(8-6-17)24(3,4)20-11-15-22(26)16-12-20/h5-16,25-26H,1-4H3

InChI key

GIXXQTYGFOHYPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sangwoo Lee et al.
Toxicology in vitro : an international journal published in association with BIBRA, 40, 297-304 (2017-02-09)
As adverse health effects of bisphenol A (BPA) become a growing public health concern, the chemicals substituting BPA have been increasingly used in everyday lives. BPA substitutes have been frequently detected in both environment and biota in increasing levels. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service