Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94821

Supelco

Guaiaverin

analytical standard

Synonym(s):

Quercetin 3-O-α-L-arabinopyranoside, 3′,4′,5,7-Tetrahydroxyflavone 3-O-α-L-arabinoside, 3-(α-L-Arabinopyranosyloxy)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, Feniculin, Foeniculin, Guaijaverin, Quercetin 3-O-α-L-arabinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O11
CAS Number:
Molecular Weight:
434.35
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤12.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1CO[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@H]1O

InChI

1S/C20H18O11/c21-8-4-11(24)14-13(5-8)30-18(7-1-2-9(22)10(23)3-7)19(16(14)27)31-20-17(28)15(26)12(25)6-29-20/h1-5,12,15,17,20-26,28H,6H2/t12-,15-,17+,20-/m0/s1

InChI key

PZZRDJXEMZMZFD-IEGSVRCHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Guaiaverin belongs to the class of flavonoids present in a wide variety of plants, which are of high research interest due to their attractive biological functions, ubiquitous nature and negligible toxicity.

Application

Guaiaverin may be used as an analytical standard for the determination of the analyte in plants, plant extracts, flowers and flower extracts, and nutraceuticals by high performance liquid chromatography (HPLC) based techniques.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Marchelak et al.
Journal of pharmaceutical and biomedical analysis, 184, 113121-113121 (2020-03-01)
The paper presents development and validation of a RP-HPLC-PDA method for quantification of 30 phenolic constituents of the blackthorn (Prunus spinosa L.) flower, a traditional European herbal medicine with a unique and complex composition. The target analytes were selected from
Wei Jia et al.
Journal of chromatography. A, 1608, 460408-460408 (2019-08-06)
Antioxidant nutraceuticals functional characteristic science is a challenging field for combining sensitivity and comprehensiveness. A untargeted screening and quantification method based on ultra-high performance liquid chromatography coupled to Quadrupole-Orbitrap high resolution mass spectrometry has been developed for determination of multiple
Martina Blunder et al.
Food chemistry, 218, 600-609 (2016-10-11)
We present a standardized, straightforward and efficient approach applicable in routine analysis of flavonoids combining sensitive NMR and HPLC experiments. The determination of the relative configuration of sugar moieties usually requires the acquisition of 13C NMR shift values. We use
Identification of flavonoids in plants by HPLC
Moiseev, et al.
Pharmaceutical Chemistry Journal, 45, 47-50 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service