Skip to Content
MilliporeSigma
All Photos(2)

Documents

94474

Supelco

D-(+)-Sucrose octaacetate

analytical standard

Synonym(s):

Sucrose octaacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38O19
CAS Number:
Molecular Weight:
678.59
Beilstein:
79290
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (TLC)

optical activity

[α]/D 58±3°, c = 2.5 in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

260 °C (lit.)

mp

82-85 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)OC[C@H]1O[C@H](O[C@]2(COC(C)=O)O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@@H]2OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C28H38O19/c1-12(29)37-9-20-22(40-15(4)32)24(42-17(6)34)25(43-18(7)35)27(45-20)47-28(11-39-14(3)31)26(44-19(8)36)23(41-16(5)33)21(46-28)10-38-13(2)30/h20-27H,9-11H2,1-8H3/t20-,21-,22-,23-,24+,25-,26+,27-,28+/m1/s1

InChI key

ZIJKGAXBCRWEOL-SAXBRCJISA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

D-(+)-Sucrose octaacetate may be used as a substrate for the determination of formaldehyde in solution using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gas chromatographic determination of formaldehyde by sucrose octa-acetate-Columpak T
Mann.S.R and Hahn.W.K
Analytical Chemistry, 39(11), 1314-1315 (1967)
A Sclafani et al.
The American journal of physiology, 270(6 Pt 2), R1197-R1202 (1996-06-01)
Rats offered a carbohydrate solution (sugar or polysaccharide) in addition to chow typically overeat and gain excessive weight. The present study sought to determine if the palatable taste of these solutions contributes to the overeating response. Adult female rats were
Baby Chandrika et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(21), 6266-6271 (2005-08-04)
High pressure (HP), high resolution (HR), proton nuclear magnetic resonance (1H NMR) spectroscopy has been utilized for the first time to investigate the solution structure of a carbohydrate based system, sucrose octaacetate (SOA), in supercritical CO2. The studies indicate that
Michael L Hurrey et al.
Langmuir : the ACS journal of surfaces and colloids, 22(17), 7324-7330 (2006-08-09)
The present study utilizes vibrational sum frequency generation (SFG) spectroscopy to study changes in the surface crystallinity of various peracetylated sugars, a class of materials that have a high affinity for carbon dioxide (CO(2)). Studies of the solid-air interface of
Meriel L Harwood et al.
Journal of food science, 77(10), S390-S393 (2012-08-29)
Classical detection thresholds do not predict liking, as they focus on the presence or absence of a sensation. Recently however, Prescott and colleagues described a new method, the rejection threshold, where a series of forced choice preference tasks are used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service