Skip to Content
MilliporeSigma
All Photos(1)

Documents

94450

Supelco

TOC standard

according to ISO/CEN EN1484, 50 mg/L±2 mg/L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

according to ISO/CEN EN1484

quality

stabilized

shelf life

limited shelf life, expiry date on the label

concentration

50 mg/L±2 mg/L

technique(s)

TOC: suitable

suitability

in accordance for application

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Total organic carbon (TOC) is a measure of the total amount of carbon in an organic compound. It is a critical parameter to evaluate the water quality for possible organic contamination.

Application

This analytical standard is intended for the calibration and validation of analytical instruments used to measure the total organic carbon (TOC) of drinking water, groundwater, surface water, seawater, and wastewater in accordance with ISO/CEN EN1484.

TOC standard may be used as a calibration standard in the analysis of the organic matter in water samples using ultraviolet-visible spectrophotometry technique.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation and Decay of Disinfection By-products in the Distribution System (2006)
Formation and Decay of Disinfection By-products in the Distribution System (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service