Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93867

Supelco

O-Desmethylvenlafaxine

analytical standard

Synonym(s):

4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, Desvenlafaxine

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$184.00

$184.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$184.00

About This Item

Empirical Formula (Hill Notation):
C16H25NO2
CAS Number:
Molecular Weight:
263.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$184.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water (calc. from elemental analysis)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

N(CC(C2(CCCCC2)O)c1ccc(cc1)O)(C)C

InChI

1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3

InChI key

KYYIDSXMWOZKMP-UHFFFAOYSA-N

Gene Information

General description

O-Desmethylvenlafaxine is a major active metabolite of the drug venlafaxine.[1]

Application

O-Desmethylvenlafaxine may be used as a reference standard for the determination of the analyte in biological fluids using liquid-liquid extraction followed by high-performance liquid chromatography (HPLC).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytochrome P450 2D6: Structure, Function, Regulation and Polymorphism (2016)
A high-performance liquid chromatographic method for the simultaneous determination of venlafaxine and O-desmethylvenlafaxine in biological fluids.
RDH, et al.
Therapeutic Drug Monitoring, 16(1), 100-107 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service