Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

936995

Sigma-Aldrich

MorDalPhos Pd G4 ChemBeads

new

Synonym(s):

[4-[2-[Bis(tricyclo[3.3.1.13,7]dec-1-yl)phosphino-κP]phenyl]morpholine](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H57N2O4PPdS
CAS Number:
Molecular Weight:
847.39
UNSPSC Code:
12352100
Pricing and availability is not currently available.

form

solid

Quality Level

composition

, 4-6 wt. % (loading)

reaction suitability

reagent type: catalyst

SMILES string

CNC1=CC=CC=C1C2=C([Pd]OS(C)(=O)=O)C=CC=C2.COCCNC3=CC=CC=C3P([C@]45C[C@H]6C[C@H](C[C@H](C6)C5)C4)[C@]78C[C@H]9C[C@H](C[C@H](C9)C8)C7

InChI

1S/C30H42NOP.C13H12N.CH4O3S.Pd/c1-2-4-28(27(3-1)31-5-7-32-8-6-31)33(29-15-21-9-22(16-29)11-23(10-21)17-29)30-18-24-12-25(19-30)14-26(13-24)20-30;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h1-4,21-26H,5-20H2;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;;;+1/p-1/t21-,22+,23-,24-,25?,26?,29-,30-,33?;;;

InChI key

VWMHZTMMTNRWLL-WPZLEGKQSA-M

General description

The ChemBeads product of the powerful MorDalPhos ligand for classic cross-coupling reactions combined with the Buchwald Fourth Generation Palladacycle. Bench stable, soluble in most organic solvents. Loaded on glass beads for use in high-throughput expermentation (HTE).

Application

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
For general uses, product is also available in powdered form (900276 )

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan T Ingoglia et al.
Tetrahedron, 75(32), 4199-4211 (2020-01-04)
Over the past three decades, Pd-catalyzed cross-coupling reactions have become a mainstay of organic synthesis. In particular, catalysts derived from biaryl monophosphines have shown wide utility in forming C-N bonds under mild reaction conditions. This work summarizes a variety of
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service