Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93667

Millipore

Heptakis-(2,3,6-tri-O-benzoyl)-β-cyclodextrin

suitable for GC

Synonym(s):

2,3,6-Tri-O-benzoyl-β-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C189H154O56
CAS Number:
Molecular Weight:
3321.21
MDL number:
UNSPSC Code:
12161700

Assay

≥98.0% (TLC)

technique(s)

gas chromatography (GC): suitable

Other Notes

Chiral solvating agent for 1H-NMR spectroscopy

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B.G.Uccello et al.
European Journal of Organic Chemistry, 2009-2009 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service