Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93665

Supelco

Saxitoxin dihydrochloride solution

20 μg/g in hydrochloric acid (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3aS,4R,10aS)-2,6-Diamino-4-{[(aminocarbonyl)oxy]methyl}-3a,4,8,9-tetrahydro-1H,10H-pyrrolo[1,2-c]purine-10,10-diol dihydrochloride, STX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N7O4 · 2HCl
CAS Number:
Molecular Weight:
372.21
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product 93665 is not currently sold in your country. Contact Technical Service

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g in hydrochloric acid (nominal concentration)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]3(NC(N2)=N)N(CCC3(O)O)C(N[C@H]1COC(N)=O)=N

InChI

1S/C10H17N7O4.2ClH/c11-6-15-5-4(3-21-8(13)18)14-7(12)17-2-1-9(19,20)10(5,17)16-6;;/h4-5,19-20H,1-3H2,(H2,12,14)(H2,13,18)(H3,11,15,16);2*1H

InChI key

YHAHUGQQOBPXOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Paralytic Shellfish Toxin PST for the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Analysis Note

20 μg/g in 66.3 μmol/l HCl in water

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service