Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93574

Supelco

4-Methylaminoantipyrine monohydrate

analytical standard

Synonym(s):

1,2-Dihydro-1,5-dimethyl-4-(methylamino)-2-phenyl-3H-pyrazol-3-one monohydrate, N-Methylaminophenazone monohydrate, Noramidopyrine monohydrate, Noraminopyrine monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O · H2O
CAS Number:
Molecular Weight:
235.28
Beilstein:
195234
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

7.65% water (theory, monohydrate)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O=C1C(NC)=C(C)N(C)N1C2=CC=CC=C2.O

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pedro M P Santos et al.
European journal of medicinal chemistry, 45(6), 2258-2264 (2010-02-23)
The pyrazolone derivatives antipyrine and 4-(N,N-dimethyl)-aminoantipyrine (aminopyrine) have long been used as analgesic, antipyretic and anti-inflammatory drugs. However, in spite of its recognized therapeutic benefits, the use of pyrazolones has been associated with agranulocytosis. Though the oxidation of aminopyrine by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service