Skip to Content
MilliporeSigma
All Photos(1)

Documents

93045

Sigma-Aldrich

Fluorotriphenylsilane

purum, ≥97.0% (GC)

Synonym(s):

Triphenylfluorosilane, Triphenylsilyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SiF
CAS Number:
Molecular Weight:
278.40
Beilstein:
2941368
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

purum

Assay

≥97.0% (GC)

mp

62-64 °C (lit.)

SMILES string

F[Si](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15FSi/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

UBGXLEFOIVWVRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service