Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92788

Sigma-Aldrich

(3-Trimethylsilyl-2-propynyl)triphenylphosphonium bromide

technical, ≥95% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26BrPSi
CAS Number:
Molecular Weight:
453.43
Beilstein:
3581358
MDL number:
UNSPSC Code:
12352000

grade

technical

Assay

≥95% (NT)

mp

166-167 °C (dec.) (lit.)

SMILES string

[Br-].C[Si](C)(C)C#CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C24H26PSi.BrH/c1-26(2,3)21-13-20-25(22-14-7-4-8-15-22,23-16-9-5-10-17-23)24-18-11-6-12-19-24;/h4-12,14-19H,20H2,1-3H3;1H/q+1;/p-1

InChI key

PBSHVEOONSKWJF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Horner-Wadsworth-Emmons olefination
  • Aromatic ring umpolung
  • Stereoselective Wittig reaction
  • Bromoetherification reactions
  • Olefination of Garner′s aldehyde
  • Regioselective Wittig reaction

Other Notes

Wittig reagent for the synthesis of en-ynes; the (E)-olefin is formed preferentially

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Pontikis et al.
Canadian Journal of Chemistry, 67, 2240-2240 (1989)
Convergent, enantiospecific total synthesis of the hypocholesterolemic agent (+)-compactin.
P A Grieco et al.
Journal of the American Chemical Society, 108(19), 5908-5919 (1986-09-01)
E.J. Corey et al.
Tetrahedron Letters, 1495-1495 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service