Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92397

Supelco

2,3-Dichlorophenoxyacetic acid

PESTANAL®, analytical standard

Synonym(s):

2,3-D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6Cl2O3
CAS Number:
Molecular Weight:
221.04
Beilstein:
1963475
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

170-175 °C

format

neat

SMILES string

OC(=O)COc1cccc(Cl)c1Cl

InChI

1S/C8H6Cl2O3/c9-5-2-1-3-6(8(5)10)13-4-7(11)12/h1-3H,4H2,(H,11,12)

InChI key

RBJIGQRZLITQJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dichlorophenoxyacetic acid (2,3-D) is a growth inactive auxin analog of 2,4-dichlorophenoxyacetic acid (2,4-D).

Application

2,3-Dichlorophenoxyacetic acid may be used as an internal standard in the following:
  • Analysis of 2,4-dichlorophenoxyacetic acid and 2-methyl-4-chlorophenoxyacetic acid in human urine samples using high-performance liquid chromatography coupled to diode array detector (HPLC-DAD) and gas chromatography with electron capture detection (GC-ECD).
  • Determination of acidic pharmaceuticals and endocrine disrupting compounds in untreated wastewaters and spring waters using gas chromatography coupled to mass spectrometry (GC-MS).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of 2, 4-dichlorophenoxyacetic acid and 2-methyl-4-chloro-phenoxyacetic acid in human urine
Pesticide Biochemistry and Physiology, 91-103 (2006)
Oxidation of NADH by intact segments of soybean hypocotyls and stimulation by 2, 4-D
Hicks C, et al.
Biochimica et Biophysica Acta - Biomembranes, 1375(1-2), 1-5 (1998)
Determination of acidic pharmaceuticals and potential endocrine disrupting compounds in wastewaters and spring waters by selective elution and analysis by gas chromatography--mass spectrometry
Gibson R, et al.
Journal of Chromatography A, 1169(1-2), 31-39 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service