Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92008

Millipore

Tryptone Bile Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

TBA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
bile salts mixture, 1.5 g/L
casein enzymic hydrolysate, 20.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.2±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

Escherichia coli
coliforms

Application

Tryptone Bile Agar is used for rapid detection and enumeration of Escherichia coli in foods using a modified direct plating method.

Preparation Note

Suspend 36.5 grams in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Mix well and pour into sterile Petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Károly Jambrovics et al.
Cancers, 12(3) (2020-03-15)
Randomized trials in acute promyelocytic leukemia patients have shown that treatment with a combination of all-trans retinoic acid (ATRA) and arsenic trioxide (ATO) is superior in efficacy to monotherapy, with significantly decreased mortality. So far, there are little data available
Ting-Fang Lee et al.
Biochimica et biophysica acta. Molecular cell research, 1868(2), 118891-118891 (2020-10-24)
Lung cancer is the leading cause of cancer death, and therefore the discovery of novel therapeutic targets is crucial. P21-activated kinase (PAK1) is an important oncogene involved in the signaling of actin cytoskeleton organization. Although PAK1 inhibition has been shown
Laura M Butkovich et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(39), 7559-7576 (2020-09-02)
Degeneration of locus ceruleus (LC) neurons and dysregulation of noradrenergic signaling are ubiquitous features of Parkinson's disease (PD). The LC is among the first brain regions affected by α-synuclein (asyn) pathology, yet how asyn affects these neurons remains unclear. LC-derived
Lei Shi et al.
The Journal of eukaryotic microbiology, 67(5), 521-531 (2020-05-06)
Intraflagellar transport (IFT) represents a bidirectional dynamic process that carries cargo essential for cilia building and the maintenance of ciliary function, which is important for the locomotion of single cells, intracellular and intercellular signalling transduction. Accumulated evidence has revealed that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service