Skip to Content
MilliporeSigma
All Photos(1)

Documents

91882

Sigma-Aldrich

4-(Trifluoromethyl)umbelliferyl phosphate disodium salt

for fluorescence, ≥90% (HPCE)

Synonym(s):

Disodium 4-(trifluoromethyl)umbelliferyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4F3Na2O6P
Molecular Weight:
354.08
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥90% (HPCE)

solubility

DMSO: soluble
H2O: soluble

fluorescence

λex 410 nm; λem 503 nm in 0.1 M Tris pH 9.0 (alkaline phosphatase)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[O-]P([O-])(=O)Oc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

Application

suitable as fluorogenic substrate for alkaline phosphatase

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service