Skip to Content
MilliporeSigma
All Photos(2)

Documents

916250

Sigma-Aldrich

[(IMes)AuCl]

Umicore

Synonym(s):

Chloro[1,3-bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]gold(I), 1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene gold(I) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24AuClN2
CAS Number:
Molecular Weight:
536.85
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

Quality Level

form

powder

mp

>300 °C
>300 °C

SMILES string

Cl[Au].Cc1cc(C)c(N2[C]N(C=C2)c3c(C)cc(C)cc3C)c(C)c1

InChI

1S/C21H24N2.Au.ClH/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;;/h7-12H,1-6H3;;1H/q;+1;/p-1

InChI key

YFUIKJIITPMUAT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

[(IMes)AuCl] is a homogeneous catalyst useful for cyclopropanation or carboxylation.

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service