Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91594

Supelco

N1-Acetylsulfamethoxazole

analytical standard

Synonym(s):

N-[(4-Aminophenyl)sulfonyl]-N-(5-methyl-3-isoxazolyl)acetamide, N1-Acetyl-N1-(5-methyl-3-isoxazolyl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N3O4S
CAS Number:
Molecular Weight:
295.31
Beilstein:
893012
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

NC1=CC=C(S(=O)(N(C2=NOC(C)=C2)C(C)=O)=O)C=C1

InChI

1S/C12H13N3O4S/c1-8-7-12(14-19-8)15(9(2)16)20(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3

InChI key

WOTJLXVSBKKTTH-UHFFFAOYSA-N

General description

N1-Acetylsulfamethoxazole is a metabolite of the bacteriostatic antibiotic sulfamethoxazole, which is typically utilized for the treatment of various diseases in humans as well as animals.

Application

N1-Acetylsulfamethoxazole may be used as an analytical standard for the determination of the analyte in soil and plant samples by liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Blasioli et al.
Journal of colloid and interface science, 419, 148-159 (2014-02-05)
Sulfonamide antibiotics are persistent pollutants present in surface and subsurface waters in both agricultural and urban environments. Sulfonamides are of particular concern in the environment because they are known to induce high levels of bacterial resistance. Adsorption of sulfamethoxazole sulfonamide
Renato F Dantas et al.
Journal of hazardous materials, 150(3), 790-794 (2007-06-19)
Sulfamethoxazole (SMX) is a bacteriostatic antibiotic largely used for diverse types of illness. Its widely use in humans and even in animals releases unmetabolized and active metabolites that have a strong potential in terms of effect in organisms. In this
Uptake, translocation and transformation of three pharmaceuticals in green pea plants
Klement A, et al.
Journal of Hydrology and Hydromechanics, 68(1) (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service