Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

909580

Sigma-Aldrich

TIPS-5-Ethynyl-dU-CEP

Synonym(s):

TIPS-5-EdU CEP, TIPS-5-Ethynyl dU CEP, TIPS-5-Ethynyl dU phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H67N4O8PSi
Molecular Weight:
911.15
UNSPSC Code:
12352200

form

powder

reaction suitability

reaction type: click chemistry

mp

>300 °C

storage temp.

−20°C

Application

This functional phosphoramidite is used to introduce click handles universally (5′ internal and 3′ end) into oligonucleotides under standard conditions for solid phase synthesis. Afterwards the oligonucleotide (RNA, PNA, DNA) can be efficiently conjugated using a copper catalyzed click reaction to azide functionalized moieties such as fluorescent dye azides and azido amino acids. In contrast to 5-Ethynyl dU CEP, TIPS-5-EdU CEP allows side product free synthesis of highly modified oligonucleotides (>10 modifications).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachin A Ingale et al.
The Journal of organic chemistry, 78(22), 11271-11282 (2013-10-22)
Clickable oligonucleotides with ethynyl residues in the 5-position of pyrimidines ((eth)dC and (eth)dU) or the 7-position of 7-deazaguanine ((eth)c(7)G(d)) are hydrated during solid-phase oligonucleotide synthesis and workup conditions. The side products were identified as acetyl derivatives by MALDI-TOF mass spectra
Johannes Gierlich et al.
Organic letters, 8(17), 3639-3642 (2006-08-11)
[reaction: see text] We report the development of the Cu(I)-catalyzed Huisgen cycloaddition (click) reaction for the multiple postsynthetic labeling of alkyne-modified DNA. A series of alkyne-modified oligodeoxyribonucleotides (ODNs) of increasing alkyne density were prepared, and the click reaction using various

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service