Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

909491

Sigma-Aldrich

4-Azidobenzoic acid

≥97%

Synonym(s):

4-N3-benzoic acid, 4-Azido-benzoic acid, P-Azidobenzoic acid, Benzoic acid, 4-azido-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
MDL number:
UNSPSC Code:
12352200

Assay

≥97%

form

powder

availability

available only in USA

mp

175 °C (dec.) (lit.)

storage temp.

2-8°C

InChI

1S/C7H5N3O2/c8-10-9-6-3-1-5(2-4-6)7(11)12/h1-4H,(H,11,12)

InChI key

PQXPAFTXDVNANI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Azidobenzoic acid is an aromatic azide used in copper(I)-catalyzed azide-alkyne cycloaddition reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-react. B - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Blood, Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Xie et al.
Bioorganic & medicinal chemistry, 15(1), 458-473 (2006-10-19)
Protein tyrosine phosphatases (PTPs) are important regulators of signal transduction pathways. Potent and selective PTP inhibitors are useful for probing these pathways and also may serve as drugs for the treatment of a variety of diseases including type 2 diabetes
Fiona Rask et al.
Journal of biomedical materials research. Part A, 95(1), 105-117 (2010-06-12)
Myocardial infarction (MI) results in the death of cardiomyocytes (CM), which causes scar formation and pathological remodeling of the heart. The delivery of healthy myocytes or bone marrow cells reduces pathological remodeling after MI, however, current cell injection methods have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service