Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90804

Sigma-Aldrich

Tributylmethylammonium iodide

purum, ≥99.0% (AT)

Synonym(s):

Methyltributylammonium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)3N(I)CH3
CAS Number:
Molecular Weight:
327.29
Beilstein:
3567734
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥99.0% (AT)

mp

184-185 °C

solubility

H2O: 0.1 g/mL, clear, colorless

SMILES string

[I-].CCCC[N+](C)(CCCC)CCCC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service