Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

907421

Sigma-Aldrich

(2R,4aR,6R,7S,8R,8aR)-7-Hydroxy-2-phenyl-6-(phenylthio)hexahydropyrano[3,2-d][1,3]dioxin-8-yl 4-oxopentanoate

≥95%

Synonym(s):

Phenyl 3-O-levulinoyl-4,6-O-benzylidine-1-thio-α-D-mannoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26O7S
Molecular Weight:
458.52
UNSPSC Code:
12352005

Assay

≥95%

form

powder

mp

127-128 °C

storage temp.

2-8°C

Application

This is a protected mannopyranoside useful as a building block for synthesis of complex carbohydrates. The compound has α-phenylthio, 3-levulinlyl, and 4,6-benzylidene protecting groups.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbohydrates and their biologically active glycoconjugate analogs are emerging as an important class of biomolecules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service