Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90668

Supelco

Phenyl 4-hydroxybenzoate

analytical standard

Synonym(s):

4-(Phenoxycarbonyl)phenol, 4-Hydroxybenzoic acid phenyl ester, Phenyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O3
CAS Number:
Molecular Weight:
214.22
Beilstein:
2645948
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

format

neat

storage temp.

2-8°C

InChI

1S/C13H10O3/c14-11-8-6-10(7-9-11)13(15)16-12-4-2-1-3-5-12/h1-9,14H

InChI key

GJLNWLVPAHNBQN-UHFFFAOYSA-N

General description

Phenyl 4-hydroxybenzoate is commonly used as a preservative in cosmetic products and is to be used within a certain concentration range as permitted by the EU regulation (EC) No 1223/2009.

Application

Phenyl 4-hydroxybenzoate may be used as an analytical reference standard for the quantification of the analyte in cosmetics using high-performance liquid chromatography equipped with photodiode array detection-electrospray ionization-mass spectrometry (HPLC-PDA-ESI/MS). It may also be used as an internal standard for the determination of thiosulfate in biological fluids using liquid chromatography-tandem mass spectrometry (LC-MS/MS) technique.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid chromatography-tandem mass spectrometry method for the determination of thiosulfate in human blood and urine as an indicator of hydrogen sulfide poisoning
Maseda C, et al.
Legal medicine, 24, 67-74 (2017)
Screening of preservatives by HPLC-PDA-ESI/MS: A focus on both allowed and recently forbidden compounds in the new EU cosmetics regulation
Lecce R, et al.
Journal of Pharmaceutical and Biomedical Analysis, 125, 260-269 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service