Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90533

Supelco

(+)-Dihydrocinchonine

analytical standard

Synonym(s):

Hydrocinchonine, Dihydrocinchonine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2O
CAS Number:
Molecular Weight:
296.41
Beilstein:
4296749
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)
95.0-105.0% (NT)

optical activity

[α]/D 197±4°, c = 0.25 in ethanol

shelf life

limited shelf life, expiry date on the label

mp

269-272 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

CC[C@H]1CN2CC[C@H]1C[C@@H]2[C@@H](O)c3ccnc4ccccc34

InChI

1S/C19H24N2O/c1-2-13-12-21-10-8-14(13)11-18(21)19(22)16-7-9-20-17-6-4-3-5-15(16)17/h3-7,9,13-14,18-19,22H,2,8,10-12H2,1H3/t13-,14-,18+,19-/m0/s1

InChI key

WFJNHVWTKZUUTR-QAMTZSDWSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service