Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90336

Supelco

Sebacic acid bis(N-succinimidyl) ester

≥97.0%

Synonym(s):

DSSeb, Di(N-succinimidyl) sebacate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N2O8
CAS Number:
Molecular Weight:
396.39
Beilstein:
6880740
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

Quality Level

Assay

≥97.0% (CHN)
≥97.0%

form

powder

reaction suitability

reagent type: cross-linking reagent

mp

162-167 °C

storage temp.

−20°C

SMILES string

O=C(CCCCCCCCC(=O)ON1C(=O)CCC1=O)ON2C(=O)CCC2=O

InChI

1S/C18H24N2O8/c21-13-9-10-14(22)19(13)27-17(25)7-5-3-1-2-4-6-8-18(26)28-20-15(23)11-12-16(20)24/h1-12H2

InChI key

XPKIVYVYXKPGBP-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Reagent for crosslining peptides and proteins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service