Skip to Content
MilliporeSigma
All Photos(2)

Documents

901995

Sigma-Aldrich

Diazo biotin-alkyne

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H53N7O9S
CAS Number:
Molecular Weight:
795.94
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

≥90%

form

solid

storage temp.

−20°C

Application

Diazo biotin-alkyne is an alkyne-activated, sodium dithionite (Na2S2O4) cleavable biotin probe that allows for efficient release of the captured biomolecules from streptavidin. This reagent contains a biotin moiety linked to an azide group through a spacer arm containing a diazo group that can be cleaved with 50 mM sodium dithionite solution.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service