Skip to Content
MilliporeSigma
All Photos(1)

Documents

901981

Sigma-Aldrich

Methyltetrazine-amine hydrochloride

Synonym(s):

(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)methanamine HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClN5
CAS Number:
Molecular Weight:
237.69
UNSPSC Code:
12352106

form

powder or crystals

availability

available only in USA

storage temp.

−20°C

Application

The stability of Methyltetrazine-amine is substantially improved compared to hydrogen substituted tetrazine-amine. Superior stability of Methyltetrazine-amine allows this reagent to be used in a wider range of chemical transformations. Long-term storage of Methyltetrazine-amine, especially in aqueous buffer, is also greatly improved compared to tetrazine-amine. This reagent is supplied as an HCl salt for improved stability and easy handling.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service