Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901760

Sigma-Aldrich

StickyCat PF6

≥95%

Synonym(s):

(1,3-Dimesityl-4-((trimethylammonio)methyl)imidazolidin-2-ylidene)dichloro(2-isopropoxybenzylidene)ruthenium(II) hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H48Cl2F6N3OPRu
CAS Number:
Molecular Weight:
843.72
UNSPSC Code:
12352101

Assay

≥95%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

266 °C (decomposition)

storage temp.

2-8°C

Application

StickyCat PF6 is for heterogeneous olefin metathesis that can be performance modulated by ion exchange. The catalyst also has high stability in non-degassed water.

Legal Information

Sold in collaboration with Apeiron Synthesis, Inc. Research quantities are 100 g / customer / year. For quantities that exceed the research limit, please contact the supplier at www.apeiron-synthesis.com. PCT/EP2013/053967

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service