Skip to Content
MilliporeSigma
All Photos(1)

Documents

90026

Supelco

Isoproturon

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3-(4-Isopropylphenyl)-1,1-dimethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O
CAS Number:
Molecular Weight:
206.28
Beilstein:
2214033
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

100
300

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

SMILES string

CC(C)c1ccc(NC(=O)N(C)C)cc1

InChI

1S/C12H18N2O/c1-9(2)10-5-7-11(8-6-10)13-12(15)14(3)4/h5-9H,1-4H3,(H,13,15)

InChI key

PUIYMUZLKQOUOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Isoproturon is a phenyl urea-derived systemic herbicide used widely for pre- and post-emergence control of annual grasses and broad-leaved weeds for wheat, barley, and rye. This selective herbicide is absorbed by roots and leaves and inhibits the electron transport in photosystem II (PS II) by binding to the D1 protein in the thylakoid membrane.

Isoproturon has been detected worldwide in soil, groundwater, surface water, or even in drinking water, exceeding the threshold value of 0.1  μg L−1, as a result, the European Union (EU) decided to ban its use in 2016. The EU has classified it as a priority micro-pollutant and has regulated its maximum residue level (MRL) at 0.3  μg L−1 for inland surface waters.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Isoprocarb CRM may also find its use as described below:
  • To elucidate the effect of isoproturon on the growth, physiological, and biochemical responses of wheat (Triticum aestivum)
  • To develop a molecularly imprinted polymer (MIP) based glassy carbon electrode for the electrochemical detection of isoproturon in water
  • To conduct a physicochemical pre-treatment for the photocatalytic destruction of metobromuron and isoproturon by different advanced oxidation processes (AOPs) in a small batch photoreactor and under direct sunlight
  • To investigate the degradation of isoproturon using Trametes versicolor laccase and its laccase-mediator systems and study the influence of the operational parameters (pH, temperature, metal ions, and PEG) on the degradation process
  • To study the effect of pH and the role of organic matter in the adsorption of isoproturon on three different types of soils and on quartz as natural adsorbents

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service