Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89793

Supelco

Fluoranthene

analytical standard

Synonym(s):

Benzo[j,k]fluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
Beilstein:
1907918
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

bp

384 °C (lit.)

mp

105-110 °C (lit.)

format

neat

SMILES string

c1ccc-2c(c1)-c3cccc4cccc-2c34

InChI

1S/C16H10/c1-2-8-13-12(7-1)14-9-3-5-11-6-4-10-15(13)16(11)14/h1-10H

InChI key

GVEPBJHOBDJJJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

388.4 °F - closed cup

Flash Point(C)

198.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service