Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

89556

Supelco

Ginkgolide J

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O10
CAS Number:
Molecular Weight:
424.40
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[H]C1[C@]2([H])OC(=O)[C@@H](C)[C@]2(O)[C@@]34OC5OC(=O)[C@H](O)C56[C@@H]([C@H](O)[C@@H](OC3=O)[C@@]146)C(C)(C)C

InChI

1S/C20H24O10/c1-6-12(23)27-7-5-17-11-8(21)9(16(2,3)4)18(17)10(22)13(24)29-15(18)30-20(17,14(25)28-11)19(6,7)26/h6-11,15,21-22,26H,5H2,1-4H3/t6-,7+,8-,9+,10+,11-,15+,17?,18+,19-,20-/m1/s1

InChI key

LMEHVEUFNRJAAV-XNSMQBOTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: ginkgo

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B N Chen et al.
Cardiovascular research, 27(9), 1576-1579 (1993-09-01)
The aim was to investigate (1) the relationship between atrial natriuretic factor (ANF) release and the extent of ischaemia-hypoxia, and (2) the potential role of eicosanoids in ANF release during global ischaemia, particularly the cyclo-oxygenase derivatives (prostaglandins) and the lipoxygenase
Jianping Zhao et al.
Acta crystallographica. Section C, Crystal structure communications, 58(Pt 3), o195-o198 (2002-03-01)
A low-temperature structure of ginkgolide A monohydrate, (1R,3S,3aS,4R,6aR,7aR,7bR,8S,10aS,11aS)-3-(1,1-dimethylethyl)-hexahydro-4,7b-dihydroxy-8-methyl-9H-1,7a-epoxymethano-1H,6aH-cyclopenta[c]furo[2,3-b]furo[3',2':3,4]cyclopenta[1,2-d]furan-5,9,12(4H)-trione monohydrate, C(20)H(24)O(9) x H(2)O, obtained from Mo K alpha data, is a factor of three more precise than the previous room-temperature determination. A refinement of the ginkgolide A monohydrate structure with Cu
H F Wang et al.
Se pu = Chinese journal of chromatography, 18(5), 394-397 (2003-01-25)
A new rapid analytical method was developed for the quantification of terpene lactones (bilobalide(BB) and ginkgolide A, B, C, J) in extract of Ginkgo biloba L. (EGb) using a liquid-liquid solvent extraction procedure followed by high performance liquid chromatography. EGb
Ottavio Vitolo et al.
Neurobiology of aging, 30(2), 257-265 (2007-07-21)
A new Ginkgo biloba extract P8A (TTL), 70% enriched with terpene trilactones, prevents A beta(1-42) induced inhibition of long-term potentiation in the CA1 region of mouse hippocampal slices. This neuroprotective effect is attributed in large part to ginkgolide J that
Stanislav Jaracz et al.
The Journal of organic chemistry, 67(13), 4623-4626 (2002-06-22)
Protocols for selective acetylation of the hydroxyl groups of ginkgolide C have been developed. These acetylations have given rise to various ginkgolide C acetates and iso-ginkgolide C acetates, the latter having a rearranged skeleton resulting from translactonization. These acetyl derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service