Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89547

Millipore

TKT Agar

suitable for microbiology

Synonym(s):

Streptococcus agalactiae selective agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

composition

aesculin, 1 g/L
agar, 13 g/L
crystal violet, 0.0013 g/L
meat extract, 5 g/L
meat peptone, 10 g/L
sodium chloride, 5 g/L
thallium sulfate, 0.333 g/L

final pH

7.4±0.2 (37 °C)

application(s)

microbiology

Application

For the isolation of Streptococcus agalactiae, the most frequent cause of bovine mastitis.

Other Notes

Determination of S. agalactiae by means of serological methods and CAMP-test

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.R. Villanueva et al.
J. Am. Vet. Assoc., 198, 1398-1398 (1991)
J. Remmers
Archiv Fur Lebensmittelhygiene, 26, 104-104 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service