Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89531

Sigma-Aldrich

Titanium(IV) oxychloride - hydrochloric acid solution

~15% Ti basis, 38-42% HCl basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiOCl2 · (HCl)x
CAS Number:
Molecular Weight:
134.77 (free base basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

composition

HCl, 38-42%
Ti, ~15%

density

1.58 g/mL at 20 °C

SMILES string

Cl.Cl[Ti](Cl)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service