Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89255

Sigma-Aldrich

Thulium(III) trifluoromethanesulfonate

purum, ≥97.0% (KT)

Synonym(s):

Thulium(III) triflate, Trifluoromethanesulfonic acid thulium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Tm
CAS Number:
Molecular Weight:
616.14
MDL number:
UNSPSC Code:
12352300

grade

purum

Assay

≥97.0% (KT)

SMILES string

[Tm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Tm/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

PBASUZORNBYVFM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Asymmetric alpha-amination and cycloadditions
  • Benzyl etherification
  • Condensation of aldehydes, amines and vinyl ethers
  • Glycosylation

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service