Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89117

Supelco

8-Methyl-6-nonenoic acid, predominantly trans

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$384.00

$384.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$384.00

About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
Beilstein:
1704210
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$384.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥80% (GC)

shelf life

limited shelf life, expiry date on the label

concentration

≤15.0% (cis-8-Methyl-6-nonenoic acid)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

refractive index

n20/D 1.440-1.450

format

neat

SMILES string

OC(CCCC/C=C/C(C)C)=O

InChI

1S/C10H18O2/c1-9(2)7-5-3-4-6-8-10(11)12/h5,7,9H,3-4,6,8H2,1-2H3,(H,11,12)/b7-5+

InChI key

OCALSPDXYQHUHA-FNORWQNLSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service