Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89017

Sigma-Aldrich

3-Thiopheneacetic acid

technical, ≥94.0% (T)

Synonym(s):

3-Thienylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2S
CAS Number:
Molecular Weight:
142.18
Beilstein:
113622
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 89017, please contact your local MilliporeSigma office or dealer. Contact Technical Service

grade

technical

Quality Level

Assay

≥94.0% (T)

form

powder

mp

73-76 °C (lit.)
73-76 °C

functional group

carboxylic acid

SMILES string

OC(=O)Cc1ccsc1

InChI

1S/C6H6O2S/c7-6(8)3-5-1-2-9-4-5/h1-2,4H,3H2,(H,7,8)

InChI key

RCNOGGGBSSVMAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria M Pérez-Madrigal et al.
ACS applied materials & interfaces, 6(12), 9719-9732 (2014-05-27)
Nanomembranes have been prepared by spin-coating mixtures of a polythiophene (P3TMA) derivative and thermoplastic polyurethane (TPU) using 20:80, 40:60, and 60:40 TPU:P3TMA weight ratios. After structural, topographical, electrochemical, and thermal characterization, properties typically related with biomedical applications have been investigated:

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service