Skip to Content
MilliporeSigma
All Photos(1)

Documents

88335

Supelco

N-(Dimethylthexylsilyl)-dimethylamin

Selectophore, ≥99.0% (T)

Synonym(s):

N,N-Dimethylthexyldimethylsilylamine, N-(Thexyldimethylsilyl)dimethylamine, N-[(2,3-Dimethyl-2-butyl)dimethylsilyl]dimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H25NSi
CAS Number:
Molecular Weight:
187.40
MDL number:
UNSPSC Code:
26111700

product line

Selectophore

Assay

≥99.0% (T)

SMILES string

CC(C)C(C)(C)[Si](C)(C)N(C)C

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Silanizing agent for glass, used e.g. for micro-electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Szabo et al.
Helvetica Chimica Acta, 67, 2128-2128 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service