Skip to Content
MilliporeSigma
All Photos(1)

Documents

87285

Sigma-Aldrich

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide

purum, ≥97.0% (GC)

Synonym(s):

1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)-2,3,5,6-tetrafluorobenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6F4CH2Br
CAS Number:
Molecular Weight:
310.99
Beilstein:
8423469
MDL number:
UNSPSC Code:
12352100

vapor pressure

0.38 psi ( 20 °C)

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.447
n20/D 1.448 (lit.)

density

1.864 g/mL at 25 °C (lit.)

SMILES string

Fc1c(F)c(c(F)c(F)c1CBr)C(F)(F)F

InChI

1S/C8H2BrF7/c9-1-2-4(10)6(12)3(8(14,15)16)7(13)5(2)11/h1H2

InChI key

WKPYRDRWTNJBQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Derivatizing agent for use in conjunction with gas chromatography-electron-capture negative ion mass spectrometry, GC-ECNI-MS

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Saha et al.
Journal of chromatography, 641(2), 400-404 (1993-07-09)
4-(Trifluoromethyl)-2,3,5,6-tetrafluorobenzyl bromide (TTBB) was synthesized in a single step from alpha,alpha,alpha,2,3,5,6-heptafluoro-p-xylene. The purpose of TTBB is to function as an analogue of pentafluorobenzyl bromide (PFBB) in electrophoric derivatization reactions prior to detection by gas chromatography-electron-capture negative ion mass spectrometry (GC-ECNI-MS).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service