Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87263

Supelco

Tetraethylammonium benzoate

purum, ≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)4N(C6H5COO)
CAS Number:
Molecular Weight:
251.36
Beilstein:
5367653
EC Number:
MDL number:
UNSPSC Code:
26111700

grade

purum

Assay

≥97.0% (T)

SMILES string

CC[N+](CC)(CC)CC.[O-]C(=O)c1ccccc1

InChI

1S/C8H20N.C7H6O2/c1-5-9(6-2,7-3)8-4;8-7(9)6-4-2-1-3-5-6/h5-8H2,1-4H3;1-5H,(H,8,9)/q+1;/p-1

InChI key

CIFIGXMZHITUAZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service