Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87215

Supelco

Trimethyltetradecylammonium hydrogen sulfate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

Tetradecyltrimethylammonium hydrogen sulfate, Trimethyltetradecylammonium bisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(HSO4)(CH3)3
CAS Number:
Molecular Weight:
353.56
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Assay

≥99.0% (T)

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.1
λ: 290 nm Amax: 0.06
λ: 300 nm Amax: 0.04
λ: 310 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

OS([O-])(=O)=O.CCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C17H38N.H2O4S/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;1-5(2,3)4/h5-17H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

LRWSTBIEJXJNJY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Trimethyltetradecylammonium hydrogen sulfate may be used as an anionic ion pairing reagent in the quantification of non derivatized sialic acids in glycoproteins using high performance liquid chromatography coupled with nano quantity analyte detector (HPLC-NQAD) and dionex-based high-pH anion-exchange chromatography with pulsed amperometric detection (HPAEC-PAD).

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel Approach for Quantitation of Non derivatized Sialic Acid in Protein Therapeutics Using Hydrophilic Interaction Chromatographic Separation and Nano Quantity Analyte Detection.
Chemmalil L, et al.
Journal of Pharmaceutical Sciences, 104(1), 15-24 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service