Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86692

Sigma-Aldrich

Tetraethylene glycol monoheptyl ether

≥98.0% (GC)

Synonym(s):

C7E4, Heptyl tetraethylene glycol ether, Heptyltetraglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6(OCH2CH2)4OH
CAS Number:
Molecular Weight:
292.41
Beilstein:
1865961
MDL number:
UNSPSC Code:
12162002

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.448

density

0.977 g/mL at 20 °C

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O(CCOCCOCCCCCCC)CCOCCO

InChI

1S/C15H32O5/c1-2-3-4-5-6-8-17-10-12-19-14-15-20-13-11-18-9-7-16/h16H,2-15H2,1H3

InChI key

KFGBNEAUNSZVPZ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service