Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83175

Sigma-Aldrich

L-Pyroglutamic acid ethyl ester

puriss., ≥98.5% (sum of enantiomers, HPLC)

Synonym(s):

(S)-5-Oxoproline ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO3
CAS Number:
Molecular Weight:
157.17
Beilstein:
82621
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

Assay

≥98.5% (sum of enantiomers, HPLC)

optical activity

[α]20/D +3.5±0.5°, c = 10% in ethanol

optical purity

enantiomeric ratio: ≥99:1 (GC)

mp

54-56 °C

application(s)

peptide synthesis

SMILES string

CCOC(=O)[C@@H]1CCC(=O)N1

InChI

1S/C7H11NO3/c1-2-11-7(10)5-3-4-6(9)8-5/h5H,2-4H2,1H3,(H,8,9)/t5-/m0/s1

InChI key

QYJOOVQLTTVTJY-YFKPBYRVSA-N

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service