Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83173

Sigma-Aldrich

D-Pyroglutamic acid ethyl ester

puriss., ≥99.0% (sum of enantiomers, HPLC)

Synonym(s):

(R)-5-Oxoproline ethyl ester, D-5-Oxo-2-pyrrolidinecarboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO3
CAS Number:
Molecular Weight:
157.17
Beilstein:
82622
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D −4.0±1.0°, c = 10% in ethanol

optical purity

enantiomeric ratio: ≥99:1 (GC)

mp

53-57 °C

application(s)

peptide synthesis

SMILES string

CCOC(=O)[C@H]1CCC(=O)N1

InChI

1S/C7H11NO3/c1-2-11-7(10)5-3-4-6(9)8-5/h5H,2-4H2,1H3,(H,8,9)/t5-/m1/s1

InChI key

QYJOOVQLTTVTJY-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This ester of pyroglutamic acid can be transformed in many ways, e.g. N-alkylation; Thionation of the lactam; Preparation of the aldehyde; Reduction to the hydroxymethyl compound

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U. Burkard et al.
Liebigs Ann. Chem., 1030-1030 (1986)
T C Friedman et al.
Biochemistry, 24(15), 3907-3913 (1985-07-16)
Pyroglutamyl-peptide hydrolase (EC 3.4.11.8) removes the N-terminal pyroglutamyl residue from pyroglutamyl-containing peptides such as thyrotropin-releasing hormone (TRH), luteinizing hormone-releasing hormone (LH-RH), neurotensin, and bombesin. The aldehyde analogue of pyroglutamate, 5-oxoprolinal, was synthesized as an active site directed transition-state inhibitor of
T.P. Andersen et al.
Liebigs Ann. Chem., 269-269 (1986)
R.B. Silverman et al.
The Journal of Organic Chemistry, 45, 815-815 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service