Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82547

Sigma-Aldrich

(−)-Pseudoephedrine

purum, ≥96.0% (sum of enantiomers, GC)

Synonym(s):

(−)-ψ-Ephedrine, (1R,2R)-2-Methylamino-1-phenyl-1-propanol, L-Pseudoephedrine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$139.00

$139.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$139.00

About This Item

Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
Beilstein:
2803417
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$139.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

purum

Assay

≥96.0% (sum of enantiomers, GC)

form

crystals

optical activity

[α]20/D −51±2°, c = 0.6% in ethanol

mp

116-119 °C

SMILES string

CN[C@H](C)[C@H](O)c1ccccc1

InChI

1S/C10H15NO/c1-8(11-2)10(12)9-6-4-3-5-7-9/h3-8,10-12H,1-2H3/t8-,10+/m1/s1

InChI key

KWGRBVOPPLSCSI-SCZZXKLOSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service