Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82545

Sigma-Aldrich

(+)-Pseudoephedrine

purum, ≥96.0% (sum of enantiomers, GC)

Synonym(s):

(+)-ψ-Ephedrine, (1S,2S)-2-Methylamino-1-phenyl-1-propanol, d-Isoephedrine, d-Pseudoephedrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
Beilstein:
2414132
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

Assay

≥96.0% (sum of enantiomers, GC)

optical activity

[α]20/D +53±2°, c = 0.6% in ethanol

mp

116-119 °C
118-120 °C

SMILES string

CN[C@@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO/c1-8(11-2)10(12)9-6-4-3-5-7-9/h3-8,10-12H,1-2H3/t8-,10+/m0/s1

InChI key

KWGRBVOPPLSCSI-WCBMZHEXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Non-selective adrenergic agonist; decongestant

Other Notes

Sales restrictions may apply

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service