Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81409

Supelco

Polystyrene

analytical standard, for GPC, 70,000

Synonym(s):

PS

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$303.00

$303.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$303.00

About This Item

Linear Formula:
[CH2CH(C6H5)]n
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$303.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~64000
Mp ~67500
Mw ~65000

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.02

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene is a synthetic thermoplastic, which can find a wide range of applications due to its properties like low cost, rigidity and good processability.[1] It may also be used as a good support material to achieve good immobilization, which is the key factor in determining the analytical performance of an assay.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical Uses of Immobilized Biological Compounds for Detection, Medical and Industrial Uses (1988)
Weathering of Polymers (1983)
Davis.A and Sims D et al.
Weathering of Polymers (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service