Skip to Content
MilliporeSigma
All Photos(1)

Documents

81131

Supelco

Poly(acrylic acid sodium salt)

analytical standard, for GPC, 83,000

Synonym(s):

Poly(sodium acrylate), Sodium polyacrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~73700
Mp ~82900
Mw ~123000

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.67

format

neat

storage temp.

2-8°C

InChI

1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

NNMHYFLPFNGQFZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Poly(acrylic acid sodium salt) is a hydrophilic polymer, which can find wide applications in the production of consumer productslike diapers, because of its ability to absorb many more times its mass in water.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harris SE et al.
First Place Science Fair Projects for Inquisitive Kids (2005)
Gooch.WJ et al.
Biocompatible Polymeric Materials and Tourniquets for Wounds (2010)
Lei Liu et al.
ACS nano, 7(2), 1368-1378 (2013-01-04)
Hierarchical FeOOH nanostructure array films constructed by different nanosized building blocks can be synthesized at the air-water interface via a bio-inspired gas-liquid diffusion method. In this approach, poly(acrylic acid) (PAA) as a crystal growth modifier plays a crucial role in
Priscila Stona et al.
The journal of adhesive dentistry, 15(3), 221-227 (2013-04-06)
To examine the influence of 11.5% polyacrylic acid pretreatment on the interface and bond strength of self-adhesive resin cements (Maxcem Elite, RelyX Unicem, SeT) to dentin. Fifty-six third molars were randomly divided into seven groups: RelyX ARC as control (ARC)
Chunjiao Zhou et al.
Journal of nanoscience and nanotechnology, 13(7), 4627-4633 (2013-08-02)
Poly(acrylic acid) (PAA) coated-Fe3O4 superparamagnetic nano-composites were synthesized through a solvothermal technique by using cheap and environmental friendly iron salts and PAA. Each nano-composite was composed of many small primary nanocrystals. The as-synthesized products were characterized by X-ray diffraction (XRD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service