Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

80903

Pivalic anhydride

purum, ≥98.0% (GC)

Synonym(s):

2,2-Dimethylpropionic anhydride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
[(CH3)3CCO]2O
CAS Number:
Molecular Weight:
186.25
EC Number:
216-263-1
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
386552
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

purum

assay

≥98.0% (GC)

refractive index

n20/D 1.409

bp

193 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C(=O)OC(=O)C(C)(C)C

InChI

1S/C10H18O3/c1-9(2,3)7(11)13-8(12)10(4,5)6/h1-6H3

InChI key

PGZVFRAEAAXREB-UHFFFAOYSA-N

Other Notes

This product has been replaced by 143502-ALDRICH | Trimethylacetic anhydride 99%

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

162.5 °F - closed cup

flash_point_c

72.5 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isamu Shiina et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(1), 167-172 (2009-11-12)
A variety of optically active 2-hydroxyalkanoates and the corresponding 2-acyloxyalkanoates are produced by the kinetic resolution of racemic 2-hydroxyalkanoates by using achiral 2,2-diarylacetic acid with hindered carboxylic anhydrides as the coupling reagents. The combined use of diphenylacetic acid, pivalic anhydride
M Joppich et al.
Analytical biochemistry, 153(1), 159-165 (1986-02-15)
Reaction of thyroxine with ethanol and pivalic anhydride in the presence of 4-dimethylaminopyridine quantitatively forms N,O-dipivalyl thyroxine ethyl ester. Other iodothyronines react similarly and the procedure is moisture insensitive. Apparently this reaction is successful, in contrast to similar procedures reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service