Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80614

Sigma-Aldrich

Pinostrobin

≥99.0% (TLC)

Synonym(s):

(S)-2,3-Dihydro-5-hydroxy-7-methoxy-2-phenyl-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
Beilstein:
270230
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (TLC)

SMILES string

COc1cc(O)c2C(=O)C[C@H](Oc2c1)c3ccccc3

InChI

1S/C16H14O4/c1-19-11-7-12(17)16-13(18)9-14(20-15(16)8-11)10-5-3-2-4-6-10/h2-8,14,17H,9H2,1H3/t14-/m0/s1

InChI key

ORJDDOBAOGKRJV-AWEZNQCLSA-N

Biochem/physiol Actions

Elicits intense apoptotic response from cultured leukemia cells in vitro. Strongly inhibits the Ca2+ signals involved in the control of G2/M phase cell cycle progression in Saccharomyces cerevisiae. Shows potent antiviral effect against herpes simplex virus-1.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hadi Poerwono et al.
Bioorganic & medicinal chemistry letters, 20(7), 2086-2089 (2010-03-12)
Pinostrobin (5-hydroxy-7-methoxyflavanone) obtained in relatively large amounts from fingerroot (Boesenbergia pandurata) was converted to its C-6 and C-8 prenylated derivatives. The Mitsunobu reaction, europium(III)-catalyzed Claisen-Cope rearrangement, and Claisen reaction coupled with cross-metathesis were used as the key steps. Using a
J C Le Bail et al.
Cancer letters, 156(1), 37-44 (2000-06-07)
The interaction between the estrogen receptor and 5-hydroxy-7-methoxyflavanone (pinostrobin) was studied in the presence or absence of estradiol or dehydroepiandrosterone sulfate (DHEAS), respectively, using a stably transfected human breast cancer cell line (MVLN). We also evaluated its action on the
Nwet Nwet Win et al.
Journal of natural products, 70(10), 1582-1587 (2007-09-28)
The chloroform extract of rhizomes of Boesenbergia pandurata demonstrated marked preferential cytotoxicity against human pancreatic PANC-1 cancer cells in nutrient-deprived medium. Bioactivity-directed investigation of this extract yielded four new secondary metabolites, geranyl-2,4-dihydroxy-6-phenethylbenzoate ( 1), 2',4'-dihydroxy-3'-(1''-geranyl)-6'-methoxychalcone ( 2), (1' R,2' S,6'
Tan Siew Kiat et al.
Bioorganic & medicinal chemistry letters, 16(12), 3337-3340 (2006-04-20)
Boesenbergia rotunda (L.) cyclohexenyl chalcone derivatives, 4-hydroxypanduratin A and panduratin A, showed good competitive inhibitory activities towards dengue 2 virus NS3 protease with the Ki values of 21 and 25 microM, respectively, whilst those of pinostrobin and cardamonin were observed
Chavi Yenjai et al.
Bioorganic & medicinal chemistry letters, 20(9), 2821-2823 (2010-04-07)
Flavones 1-4 isolated from Kaempferia parviflora were used for structural modification. Sixteen flavonoid derivatives, including four new derivatives, were synthesized and evaluated for cytotoxicity against KB and NCI-H187 cell lines. Flavanones 2a-4a demonstrated higher cytotoxic activity than the parent compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service