Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80534

Supelco

Ethanethiol

analytical standard

Synonym(s):

Ethyl mercaptan, Mercaptan C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SH
CAS Number:
Molecular Weight:
62.13
Beilstein:
773638
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor density

2.1 (vs air)

vapor pressure

8.51 psi ( 20 °C)

Assay

≥98.5% (GC)

autoignition temp.

570 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

18.2 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤18% water

refractive index

n20/D 1.429-1.433
n20/D 1.4306 (lit.)

bp

35 °C (lit.)

density

0.839 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCS

InChI

1S/C2H6S/c1-2-3/h3H,2H2,1H3

InChI key

DNJIEGIFACGWOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-49.0 °F - closed cup

Flash Point(C)

-45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hanczkó et al.
Journal of chromatography. A, 1163(1-2), 25-42 (2007-07-04)
The main aims of this work were (a) to present the characteristics and stability of the o-phthalaldehyde (OPA)-ethanethiol (ET) derivatives of 22 amino acids, including the believed-to-be less stable OPA derivatives providing glycine, gamma-aminobutyric acid, beta-alanine, histidine, ornithine, lysine and
Esther Vega et al.
Chemosphere, 109, 113-119 (2014-05-31)
The effectiveness of different advanced oxidation processes on the treatment of a multicomponent aqueous solution containing ethyl mercaptan, dimethyl sulphide and dimethyl disulphide (0.5 mg L(-1) of each sulphur compound) was investigated with the objective to assess which one is
Lore Geldof et al.
Biomedical chromatography : BMC, 28(7), 974-985 (2014-02-06)
Anti-doping laboratories need to be aware of evolutions on the steroid market and elucidate steroid metabolism to identify markers of misuse. Owing to ethical considerations, in vivo and in vitro models are preferred to human excretion for nonpharmaceutical grade substances.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service