Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80380625-0010-P

Europium Functionalized Microspheres

F1-EU 050-0.1G-P, polystyrene, carboxylic acid functionalized, europium labeled, vial of 0.1 g, fluorescent red

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

label

europium labeled

material

polyethylene bottle
polystyrene

packaging

vial of 0.1 g

manufacturer/tradename

Estapor®

concentration

1% solids

color

fluorescent red

diameter

0.450-0.530 μm

capacity

1-10 μeq/g surface charge density

functional group

carboxylic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Estapor® is proud to launch our new Europium microspheres. Labeled with Europium, these microspheres provide several advantages over traditional fluorescent microspheres.
Significantly improve lateral flow assay sensitivity.
  • Drastically reduce background fluorescence.
  • Make assays easier to read and quantify.
  • Exhibit a much longer Stokes shift than traditional fluorescent labels.
  • Exhibit an enhanced fluorescent quantum yield facilitating a low detection limit.
  • Are functionalized with a carboxylated surface for protein conjugation.
  • Are available in three different size options.

Application

Fluorescent Microspheres for Quantitative Lateral Flow assays.

Legal Information

Estapor is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service