Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

793140

Sigma-Aldrich

Specially Denatured Alcohol

190 proof, SDA 40 (40-2), contains 0.14 % (v/v) tert-Butyl alcohol and Brucine sulfate

Synonym(s):

Ethanol, SDA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
application(s):
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Pricing and availability is not currently available.

form

liquid

contains

0.14 % (v/v) tert-Butyl alcohol and Brucine sulfate

concentration

190 proof

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Tax and license information

Permit for use required

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

5GA is in metal drum with flex spout.
5GA-PB is in a poly drum with flex spout.
3x5GA is in metal drum with flex spout.
3x5GA-PB is in a poly drum with flex spout.
55GA is in metal drum.
55GA-PB is in poly drum.

Components

99.86% (v/v) Ethanol, 190 Proof
0.14% (v/v) tert-Butyl alcohol and Brucine sulfate

Other Notes

Ordering Information:
To purchase Specially Denatured Alcohol, customers must possess an Industrial Alcohol User Permit from the Alcohol and Tobacco Tax and Trade Bureau (TTB).
  • Permit must have customer signature and date of submission to Sigma-Aldrich in addition to the original signature and date.
  • Permit must have word, "copy" handwritten or stamped.
  • Permit number must contain correct state or US designation matching ship-to address.
  • Permit address must match ship-to-address in Sigma-Aldrich system.
  • Permit only needs to be provided on first order to [email protected].
  • All orders require that a PO with the statement, "valid permit #####" be submitted to [email protected].

TTB Link

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Allenstein et al.
Soft matter, 11(25), 5053-5059 (2015-06-02)
Cells require adhesion to survive, proliferate and migrate, as well as for wound healing and many other functions. The strength of contractile cell forces on an underlying surface is a highly relevant quantity to measure the affinity of cells to
Sonia Soloneski et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(5), 834-844 (2015-03-31)
In vitro effects of the carbamates pirimicarb and zineb and their formulations Aficida® (50% pirimicarb) and Azzurro® (70% zineb), respectively, were evaluated in Chinese hamster ovary (CHO-K1) cells. Whereas the cytokinesis-blocked micronucleus cytome assay was employed to test for genotoxicity
Danielle Hagstrom et al.
Toxicological sciences : an official journal of the Society of Toxicology, 147(1), 270-285 (2015-06-28)
Traditional toxicology testing has relied on low-throughput, expensive mammalian studies; however, timely testing of the large number of environmental toxicants requires new in vitro and in vivo platforms for inexpensive medium- to high-throughput screening. Herein, we describe the suitability of the asexual
Nicholas J Niemuth et al.
Chemosphere, 135, 38-45 (2015-04-22)
The occurrence of intersex fish, where male reproductive tissues show evidence of feminization, have been found in freshwater systems around the world, indicating the potential for significant endocrine disruption across species in the ecosystem. Estrogens from birth control medications in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service